MilliporeSigma
  • Genetic, Genomic, and Imaging Approaches to Dissect the Role of Polycomb Group Epigenetic Regulators in Mice.
Genetic, Genomic, and Imaging Approaches to Dissect the Role of Polycomb Group Epigenetic Regulators in Mice.
Methods in molecular biology (Clifton, N.J.) (2022-06-23)
Nayuta Yakushiji-Kaminatsui, Takashi Kondo, Yasuhide Ohinata, Junichiro Takano, Haruhiko Koseki
ABSTRACT
Among the most important histone methyltransferases for metazoan development are EZH1/2 and their homologs, which methylate histone H3 lysine 27 and act as part of a highly conserved set of chromatin regulators called Polycomb Group (PcG) proteins. Reaching a precise understanding of the roles of PcG proteins in the orchestration of differentiation and the maintenance of cell identity requires a variety of genetic and molecular approaches. Here, we present a full suite of methods for the study of PcG proteins in early murine development, including mutant strain generation, embryonic stem cell derivation, epigenomic profiling, and immunofluorescence and in situ hybridization.
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, This Human plasma fibronectin is a purified protein, used as an attachment factor suitable for cell propagation in vitro.
Roche
Bovine Serum Albumin, blood typing: suitable, Endonuclease, Exonuclease, Rnases, Proteinases free
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A