MilliporeSigma
  • Endosomal-associated RFFL facilitates mitochondrial clearance by enhancing PRKN/parkin recruitment to mitochondria.
Endosomal-associated RFFL facilitates mitochondrial clearance by enhancing PRKN/parkin recruitment to mitochondria.
Autophagy (2022-04-05)
Rishith Ravindran, Anoop Kumar G Velikkakath, Nikhil Dev Narendradev, Aneesh Chandrasekharan, T R Santhoshkumar, Srinivasa M Srinivasula
ABSTRACT
Mutations in the ubiquitin ligase PRKN (parkin RBR E3 ubiquitin protein ligase) are associated with Parkinson disease and defective mitophagy. Conceptually, PRKN-dependent mitophagy is classified into two phases: 1. PRKN recruits to and ubiquitinates mitochondrial proteins; 2. formation of phagophore membrane, sequestering mitochondria for degradation. Recently, endosomal machineries are reported to contribute to the later stage for membrane assembly. We reported a role for endosomes in the events upstream of phase 1. We demonstrate that the endosomal ubiquitin ligase RFFL (ring finger and FYVE like domain containing E3 ubiquitin protein ligase) associated with damaged mitochondria, and this association preceded that of PRKN. RFFL interacted with PRKN, and stable recruitment of PRKN to damaged mitochondria was substantially reduced in RFFL KO cells. Our study unraveled a novel role of endosomes in modulating upstream pathways of PRKN-dependent mitophagy initiation.Abbreviations CCCP: carbonyl cyanide 3-chlorophenylhydrazone; DMSO: dimethyl sulfoxide; EGFP: enhanced green fluorescence protein; KO: knockout; PRKN: parkin RBR E3 ubiquitin protein ligase; RFFL: ring finger and FYVE like domain containing E3 ubiquitin protein ligase; UQCRC1: ubiquinol-cytochrome c reductase core protein 1; WT: wild-type.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)