MilliporeSigma
Expansion spatial transcriptomics.
Nature methods (2023-06-23)
Yuhang Fan, Žaneta Andrusivová, Yunming Wu, Chew Chai, Ludvig Larsson, Mengxiao He, Liqun Luo, Joakim Lundeberg, Bo Wang
ABSTRACT
Capture array-based spatial transcriptomics methods have been widely used to resolve gene expression in tissues; however, their spatial resolution is limited by the density of the array. Here we present expansion spatial transcriptomics to overcome this limitation by clearing and expanding tissue prior to capturing the entire polyadenylated transcriptome with an enhanced protocol. This approach enables us to achieve higher spatial resolution while retaining high library quality, which we demonstrate using mouse brain samples.
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer