MilliporeSigma
  • 1,6-hexanediol diacrylate: a rapid and potent sensitizer in the printing industry.
1,6-hexanediol diacrylate: a rapid and potent sensitizer in the printing industry.
The Australasian journal of dermatology (2000-08-24)
V A Morgan, J M Fewings
ABSTRACT
Printers are exposed to a wide variety of irritants and sensitizers. Allergy developing from a single exposure to any compound is rare. We report two cases of workers in the printing industry who developed allergic contact dermatitis to 1,6-hexanediol diacrylate (HDDA) within a short period of exposure. The first developed within weeks of exposure to a plastic sheet primed with HDDA. The second developed after a single accidental exposure.
Sigma-Aldrich
1,6-Hexanediol diacrylate, technical grade, 80%