MilliporeSigma
  • PbSe nanocrystal growth as nanocubes and nanorods on peptide nanotubes via different directed-assembly pathways.
PbSe nanocrystal growth as nanocubes and nanorods on peptide nanotubes via different directed-assembly pathways.
Nanoscale (2010-09-14)
Menglu Shi, Wei Su, Hiroshi Matsui
ABSTRACT
Pb-binding TAR-1 peptides (Ile-Ser-Leu-Leu-His-Ser-Thr) were covalently conjugated on a bolaamphiphile peptide nanotube substrate and the precursors of PbSe were incubated at room temperature. This resulted in the growth of highly crystalline PbSe nanocubes on this biomimetic cylindrical substrate. The growth mechanism to generate nanocubes occurs via the directed self-assembly of nanoparticles and then nanoparticle fusion. The peptide conformation and the cylindrical peptide nanotube substrate play important roles in the mesoscopic crystallization of PbSe nanocubes. Changing the buffer for the peptide immobilization process from 2-(N-morpholino)ethanesulfonic acid to phosphate induces a transformation in the nanocrystal shape from nanocube to nanorods. The conformational change of the TAR-1 peptide on the nanotubes due to the change in the buffer seems to be responsible for aggregating intermediate nanoparticles in different directions for the directed fusion and mesoscopic crystallization of PbSe into the different shapes.
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
SAFC
MES sodium salt