MilliporeSigma
  • Reversed-phase high-performance liquid chromatography separation of dimethylaminoazobenzene sulfonyl- and dimethylaminoazobenzene thiohydantoin-amino acid derivatives for amino acid analysis and microsequencing studies at the picomole level.
Reversed-phase high-performance liquid chromatography separation of dimethylaminoazobenzene sulfonyl- and dimethylaminoazobenzene thiohydantoin-amino acid derivatives for amino acid analysis and microsequencing studies at the picomole level.
Analytical biochemistry (1989-04-01)
V Stocchi, G Piccoli, M Magnani, F Palma, B Biagiarelli, L Cucchiarini
ABSTRACT
A simple and fast reversed-phase high-performance liquid chromatographic method has been developed for the complete separation of 35 dimethylaminoazobenzene sulfonyl (DABS)-amino acids and by-products. This method allows simultaneous determination of primary and secondary amino acids which can be present in protein and peptide hydrolysates and also detects the presence of cysteic acid, S-sulfocysteine, hydroxyproline, taurine, norleucine, cystine, and delta-hydroxylysine. The precolumn derivatization of amino acids with dimethylaminoazobenzene sulfonyl chloride (DABS-Cl) is simple and quick (10 min at 70 degrees C) and allows the complete reaction of primary and secondary amino acids. The separation of the compounds under investigation is achieved in 25 min using a reversed-phase 3-microns Supelcosil LC-18 column at room temperature. The versatility of the proposed method is documented by amino acid determination on protein samples obtained using different hydrolysis techniques (HCl, methane-sulfonic acid, and NaOH), with attention given to the detection of tryptophan in protein samples with high sugar concentration. Furthermore, we have reported the experimental conditions necessary to apply this method to the amino acid analysis of very low amount of proteins (1 to 5 micrograms) electroeluted from a stained band after sodium dodecyl sulfate-polyacrylamide gel electrophoresis. The stability of DABS-derivatives, the short time of analysis, the high reproducibility and sensitivity of the system, and the complete resolution of all compounds of interest make this method suitable for routine analysis. Furthermore, we have also developed a fast reversed-phase high-performance liquid chromatographic method for the complete separation of dimethylaminoazobenzene thiohydantoin (DABTH)-amino acids. The separation of the compounds under investigation is obtained, at room temperature, in less than 18 min using a reversed-phase Supelcosil LC-18 DB column, 3-micron particles, and also allows the complete separation of DABTH-Ile, DABTH-Leu, and DABTH-Norleu. The short time of analysis, together with the high reproducibility of the system and its sensitivity at picomole levels, make this method very suitable for the identification of DABTH-amino acids released during microsequencing studies of proteins and peptides with the dimethylaminoazobenzene isothiocyanate reagent. In addition, we have shown that it is possible to obtain complete separation of DABTH-amino acids also under isocratic conditions.(ABSTRACT TRUNCATED AT 400 WORDS)
Product Description
Supelco
SUPELCOSIL LC-18-DB Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 3 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 3 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
SUPELCOSIL LC-18-DB Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 3 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 3 μm particle size, L × I.D. 3.3 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-DB Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 30 cm × 4 mm
Sigma-Aldrich
4-(Dimethylamino)azobenzene-4′-sulfonyl chloride, ≥97.5% (AT)
Supelco
SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm