MilliporeSigma
  • Access to polyfunctionalized diquinanes, hydrindanes, and decalines via TiCl4 promoted Michael-aldol and Baylis-Hillman reactions.
Access to polyfunctionalized diquinanes, hydrindanes, and decalines via TiCl4 promoted Michael-aldol and Baylis-Hillman reactions.
Organic letters (2011-12-14)
Blandine Ressault, Alexis Jaunet, Philippe Geoffroy, Sébastien Goudedranche, Michel Miesch
ABSTRACT
The addition of 0.5 equiv of TiCl(4) to (cyclo)alkanones tethered to α,β-unsaturated ketones afforded polyfunctionalized diquinanes, hydrindanes, and decalines. These products, resulting from a Michael-aldol or a Baylis-Hillman reaction, can be obtained with high or total diastereoselectivity in moderate to high yields. These scaffolds represent interesting building blocks for the synthesis of complex natural products.
Sigma-Aldrich
cis-Decahydronaphthalene, 99%
Sigma-Aldrich
trans-Decahydronaphthalene, 99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis