MilliporeSigma
  • Synthesis and characterization of Y2O3:Er3+ upconversion materials with nanoporous structures.
Synthesis and characterization of Y2O3:Er3+ upconversion materials with nanoporous structures.
Journal of nanoscience and nanotechnology (2012-03-15)
Qipeng Lu, Yanbing Hou, Aiwei Tang, Zhihui Feng, Feng Teng, XiaoJun Liu
ABSTRACT
Y2O3:Er3+ upconversion materials with nanoporous structures were prepared by a hydrothermal method following a post-thermal treatment. The structure and morphology of the materials were characterized by X-ray diffraction (XRD) and field-emission scanning electron microscopy (FESEM). The results indicated that the as-obtained Y2O3:Er3+ powders were of cubic-phase structure, and the nanoporous structure was formed in the annealing process. The optical results indicated that high annealing temperature could improve the upconversion properties, but it could destroy the nanoporous structure. Under 980 nm excitation, red (4F(9/2) --> 4I(15/2) and green (2H(11/2), 4S(3/2) --> 4I(15/2)) upconversion luminescence was observed. The studies on the intensity dependence of upconversion emission indicated that two-photon processes were responsible for the green and red upconversion luminescence. This kind of multifunctional material has potential applications in nanocontainers for use as biomolecule and drugs carriers.
Sigma-Aldrich
Yttrium(III) oxide, nanopowder, <50 nm particle size
Sigma-Aldrich
Yttrium(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, dispersion, 10 wt. % in isopropanol, nanoparticles, <100 nm (DLS), ≥99.9% trace metals basis