MilliporeSigma
  • Universal concept for the implementation of a single cleavable unit at tunable position in functional poly(ethylene glycol)s.
Universal concept for the implementation of a single cleavable unit at tunable position in functional poly(ethylene glycol)s.
Biomacromolecules (2012-12-22)
Carsten Dingels, Sophie S Müller, Tobias Steinbach, Christine Tonhauser, Holger Frey
ABSTRACT
Poly(ethylene glycol) (PEG) with acid-sensitive moieties gained attention particularly for various biomedical applications, such as the covalent attachment of PEG (PEGylation) to protein therapeutics, the synthesis of stealth liposomes, and polymeric carriers for low-molecular-weight drugs. Cleavable PEGs are favored over their inert analogues because of superior pharmacodynamic and/or pharmacokinetic properties of their formulations. However, synthetic routes to acetal-containing PEGs published up to date either require enormous efforts or result in ill-defined materials with a lack of control over the molecular weight. Herein, we describe a novel methodology to implement a single acetaldehyde acetal in well-defined (hetero)functional poly(ethylene glycol)s with total control over its position. To underline its general applicability, a diverse set of initiators for the anionic polymerization of ethylene oxide (cholesterol, dibenzylamino ethanol, and poly(ethylene glycol) monomethyl ether (mPEG)) was modified and used to synthesize the analogous labile PEGs. The polyether bearing the cleavable lipid had a degree of polymerization of 46, was amphiphilic and exhibited a critical micelle concentration of 4.20 mg·L(-1). From dibenzylamino ethanol, three heterofunctional PEGs with different molecular weights and labile amino termini were generated. The transformation of the amino functionality into the corresponding squaric acid ester amide demonstrated the accessibility of the cleavable functional group and activated the PEG for protein PEGylation, which was exemplarily shown by the attachment to bovine serum albumin (BSA). Furthermore, turning mPEG into a macroinitiator with a cleavable hydroxyl group granted access to a well-defined poly(ethylene glycol) derivative bearing a single cleavable moiety within its backbone. All the acetal-containing PEGs and PEG/protein conjugates were proven to degrade upon acidic treatment.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)