MilliporeSigma
  • Modification of electronic properties of top-gated graphene devices by ultrathin yttrium-oxide dielectric layers.
Modification of electronic properties of top-gated graphene devices by ultrathin yttrium-oxide dielectric layers.
Nanoscale (2012-12-25)
Lin Wang, Xiaolong Chen, Yang Wang, Zefei Wu, Wei Li, Yu Han, Mingwei Zhang, Yuheng He, Chao Zhu, Kwok Kwong Fung, Ning Wang
ABSTRACT
We report the structure characterization and electronic property modification of single layer graphene (SLG) field-effect transistor (FET) devices top-gated using ultrathin Y(2)O(3) as dielectric layers. Based on the Boltzmann transport theory within variant screening, Coulomb scattering is confirmed quantitatively to be dominant in Y(2)O(3)-covered SLG and a very few short-range impurities have been introduced by Y(2)O(3). Both DC transport and AC capacitance measurements carried out at cryogenic temperatures demonstrate that the broadening of Landau levels is mainly due to the additional charged impurities and inhomogeneity of carriers induced by Y(2)O(3) layers.
Sigma-Aldrich
Yttrium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Yttrium(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, nanopowder, <50 nm particle size
Sigma-Aldrich
Yttrium(III) oxide, dispersion, 10 wt. % in isopropanol, nanoparticles, <100 nm (DLS), ≥99.9% trace metals basis
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis