MilliporeSigma
  • Preparation and use of poly(hydroxyethyl methacrylate) cryogels containing L-histidine for β-casein adsorption.
Preparation and use of poly(hydroxyethyl methacrylate) cryogels containing L-histidine for β-casein adsorption.
Journal of food science (2013-01-22)
Murat Yavuz, Zübeyde Baysal
ABSTRACT
The objective of this study was to determine β-casein adsorption by using supermacroporous poly(2-hydroxyethyl methacrylate-N-methacryloyl-(l)-histidine methyl ester) [p(HEMA-MAH)] cryogel. β-Casein adsorption properties of p(HEMA-MAH) cryogel were studied for the application of β-casein purification. The cryogel was produced by free radical polymerization initiated by N,N,N',N'-tetramethylene diamine and ammonium persulfate pairs in an ice bath. P(HEMA-MAH) cryogel was characterized by swelling tests, Fourier transform infrared spectroscopy, and scanning electron microscopy. The effects of the flow rate, pH, temperature, initial β-casein concentration, and ionic strength on the adsorption efficiency of cryogel were studied. The equilibrium swelling degree of the p(HEMA-MAH) cryogel was 6.73 g H(2) O/g cryogel. β-Casein adsorption capacity of p(HEMA-MAH) cryogel from aqueous solution was estimated as 31.17 mg/g cryogel. It was also observed that β-casein could be repeatedly adsorbed and desorbed with p(HEMA-MAH) cryogel without significant loss in the adsorption capacity.
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), BioReagent, powder, suitable for cell culture
Sigma-Aldrich
L-Histidine methyl ester dihydrochloride, 97%
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), average Mv 20,000
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), average Mv 300,000, crystalline
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), average Mv 1,000,000