MilliporeSigma
  • The effect of thermal and ultrasonic treatment on amino acid composition, radical scavenging and reducing potential of hydrolysates obtained from simulated gastrointestinal digestion of cowpea proteins.
The effect of thermal and ultrasonic treatment on amino acid composition, radical scavenging and reducing potential of hydrolysates obtained from simulated gastrointestinal digestion of cowpea proteins.
Plant foods for human nutrition (Dordrecht, Netherlands) (2013-01-29)
Joycelyn K Quansah, Chibuike C Udenigwe, Firibu K Saalia, Rickey Y Yada
ABSTRACT
The effect of thermal and ultrasonic treatment of cowpea proteins (CP) on amino acid composition, radical scavenging and reducing potential of hydrolysates (CPH) obtained from in vitro simulated gastrointestinal digestion of CP was evaluated. Hydrolysis of native and treated CP with gastrointestinal pepsin and pancreatin yielded CPH that displayed antioxidant activities based on oxygen radical scavenging capacity (ORAC), ferric reducing antioxidant power (FRAP) and superoxide radical scavenging activity (SRSA). CPH derived from the treated CP yielded higher ORAC values than CPH from untreated proteins. However, lower significant FRAP and SRSA values were observed for these samples compared to untreated CPH (p < 0.05). Amino acid analysis indicated that CP processing decreased total sulphur-containing amino acids in the hydrolysates, particularly cysteine. The amount of cysteine appeared to be positively related to FRAP and SRSA values of CPH samples, but not ORAC. The results indicated that thermal and ultrasonic processing of CP can reduce the radical scavenging and reducing potential of the enzymatic hydrolysates possibly due to the decreased amounts of cysteine. Since the hydrolysates were generated with gastrointestinal enzymes, it is possible that the resulting compounds are produced to exert some health functions during normal consumption of cowpea.
Sigma-Aldrich
Pancreatin from porcine pancreas, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, ≥3 × USP specifications
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein