MilliporeSigma
  • Aqueous synthesis of highly luminescent AgInS₂-ZnS quantum dots and their biological applications.
Aqueous synthesis of highly luminescent AgInS₂-ZnS quantum dots and their biological applications.
Nanoscale (2013-02-09)
Michelle D Regulacio, Khin Yin Win, Seong Loong Lo, Shuang-Yuan Zhang, Xinhai Zhang, Shu Wang, Ming-Yong Han, Yuangang Zheng
ABSTRACT
Highly emissive and air-stable AgInS2-ZnS quantum dots (ZAIS QDs) with quantum yields of up to 20% have been successfully synthesized directly in aqueous media in the presence of polyacrylic acid (PAA) and mercaptoacetic acid (MAA) as stabilizing and reactivity-controlling agents. The as-prepared water-dispersible ZAIS QDs are around 3 nm in size, possess the tetragonal chalcopyrite crystal structure, and exhibit long fluorescence lifetimes (>100 ns). In addition, these ZAIS QDs are found to exhibit excellent optical and colloidal stability in physiologically relevant pH values as well as very low cytotoxicity, which render them particularly suitable for biological applications. Their potential use in biological labelling of baculoviral vectors is demonstrated.
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
Sigma-Aldrich
Zinc sulfide, powder, 10 μm, 99.99% trace metals basis
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Sigma-Aldrich
Thioglycolic acid, ≥99%
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Zinc sulfide, purum, 97% (from Zn)
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
Sodium thioglycolate, ≥96.5% (iodometric)
Sigma-Aldrich
Zinc sulfide, pieces, 3-12 mm, 99.9% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
Supelco
Thioglycolic acid solution, ~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000