MilliporeSigma
  • A pH-responsive nano-carrier with mesoporous silica nanoparticles cores and poly(acrylic acid) shell-layers: fabrication, characterization and properties for controlled release of salidroside.
A pH-responsive nano-carrier with mesoporous silica nanoparticles cores and poly(acrylic acid) shell-layers: fabrication, characterization and properties for controlled release of salidroside.
International journal of pharmaceutics (2013-02-12)
Hailong Peng, Ruichen Dong, Shenqi Wang, Zhong Zhang, Mei Luo, Chunqing Bai, Qiang Zhao, Jinhua Li, Lingxin Chen, Hua Xiong
ABSTRACT
A novel pH-responsive nano-carrier MSNs-PAA, possessing mesoporous silica nanoparticles (MSNs) cores and poly(acrylic acid) (PAA) shell-layers, was developed for controlled release of salidroside. The vinyl double bonds modified MSNs were synthesized by using cetyltrimethylammonium bromide (CTAB) as templates, tetraethyl orthosilicate (TEOS) as silicon source, and 3-(trimethoxylsilyl) propyl methacrylate (MPS) as surface modification functionalities. The pH-responsive layers of PAA were grafted onto the vinyl double bonds of the MSNs via precipitation polymerization, producing the MSNs-PAA with a hollow cubic core and mesoporous shell with penetrating pore channels. The characteristic results also showed that PAA was successfully grafted onto the surface of the MSNs. The MSNs-PAA was investigated as carriers for loading and regulating the release of salidroside in different pH solutions for the first time. The results demonstrated that the PAA layers on the surface of MSNs-PAA exhibited opened and closed states at different pH values, and thus could regulate the uptake and release of salidroside. The application of such pH-responsive nano-carrier might offer a potential platform for controlled delivery and increasing the bioavailability of drugs.
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Supelco
Salidroside, analytical standard
Sigma-Aldrich
Poly(acrylic acid)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)