MilliporeSigma
  • Multifunctional magnetic-fluorescent eccentric-(concentric-Fe₃O₄@SiO₂@polyacrylic acid core-shell nanocomposites for cell imaging and pH-responsive drug delivery.
Multifunctional magnetic-fluorescent eccentric-(concentric-Fe₃O₄@SiO₂@polyacrylic acid core-shell nanocomposites for cell imaging and pH-responsive drug delivery.
Nanoscale (2013-02-12)
Lu Li, Cong Liu, Lingyu Zhang, Tingting Wang, Hong Yu, Chungang Wang, Zhongmin Su
ABSTRACT
Multifunctional fluorescent-magnetic pH-responsive eccentric-(concentric-Fe3O4@SiO2)@polyacrylic acid core-double shell nanocomposites (NCs) have been prepared for simultaneous cell imaging and pH-responsive drug delivery. To confirm the universality of the synthetic concept, the synthetic strategy was also extended to fabricate monodisperse eccentric-(concentric-NaYF4:Yb/Er/Gd@SiO2)@polyacrylic acid core-double shell NCs.
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid)
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)