MilliporeSigma
  • "Mixed" anionic and non-ionic micellar liquid chromatography for high-speed radiometabolite analysis of positron emission tomography radioligands.
"Mixed" anionic and non-ionic micellar liquid chromatography for high-speed radiometabolite analysis of positron emission tomography radioligands.
Journal of chromatography. A (2013-02-13)
Ryuji Nakao, Christer Halldin
ABSTRACT
A mixed micellar liquid chromatographic (LC) method, the mobile phase consisting of anionic and non-ionic surfactants, has been developed for the high-speed direct radiometabolite analysis of positron emission tomography (PET) radioligands in plasma. The addition of Triton X-100 on an anionic surfactant sodium dodecyl sulphate (SDS) mobile phase improved elution strength and peak efficiency for many PET radioligands. Several radioligands could be easily separated from their radioactive metabolites with short run time of only 4 min using a "pure" (without organic solvent) mixed micellar mobile phase and semi-preparative monolithic C(18)-bonded silica column by simple isocratic elution without any treatment of plasma. Moreover, the use of "hybrid" mixed micellar mobile phase containing anionic, non-ionic surfactants and organic solvent was effective to further enhance peak efficiency and elute highly retained hydrophobic PET radioligands. These characteristics enabled significant shorting the radiometabolite analysis procedure of PET radioligands and simplifying the experimental setup.
Product Description
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Triton X-102
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Tetrabenazine, ≥98% (HPLC), solid
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets