MilliporeSigma
In situ continuous growth formation of synthetic biominerals.
Chemical communications (Cambridge, England) (2013-03-20)
Natasha H Munro, David W Green, Kathryn M McGrath
ABSTRACT
Continuous self-assembled growth of both the organic and inorganic components of materials with nacre-like structure is achieved upon mineralisation of chitin and chitosan scaffolds using a combined soaking method and the inclusion of poly(acrylic acid) and chitosan oligomers as additives.
Sigma-Aldrich
Chitin from shrimp shells, BioReagent, suitable for analysis of chitinase, purified powder
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~250,000, 35 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~2,000, 50 wt. % in H2O, electronic grade
Sigma-Aldrich
Poly(acrylic acid) solution, average Mw ~100,000, 35 wt. % in H2O
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~4,000,000
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Supelco
Poly(acrylic acid), analytical standard, average Mn 130,000 (Typical)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~450,000
Sigma-Aldrich
Poly(acrylic acid)
Sigma-Aldrich
Poly(acrylic acid), average Mv ~1,250,000
Sigma-Aldrich
Poly(acrylic acid), average Mv ~3,000,000