MilliporeSigma
Catheter lubrication and fixation: interventions.
British journal of nursing (Mark Allen Publishing) (2013-06-12)
Mary Wilson
ABSTRACT
This article looks at the indications for catheterisation, the anatomy of the urethra, the importance of asepsis and the rationale for the use of urethral lubrication for catheter insertion. The variations of lubrication available are discussed, including the use of products containing lidocaine and chlorhexidine. It then considers the problems caused by traction to the catheter and discusses the securement devices available for preventing tissue damage at the bladder neck, within the urethra and also at the meatus. The variation of devices is examined along with the potential problems associated with their use. Finally, it discusses the importance of clinicians being knowledgeable about the problems looked at and how to resolve them.
Product Description
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocaine hydrochloride monohydrate, solid
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard