MilliporeSigma
  • [Effectiveness and toxicity of MOPP, ABVD, BEACOPP chemotherapy in first-diagnosed Hodgkin lymphoma with a poor prognosis].
[Effectiveness and toxicity of MOPP, ABVD, BEACOPP chemotherapy in first-diagnosed Hodgkin lymphoma with a poor prognosis].
Voprosy onkologii (2013-07-03)
L V Filatova, A A Plotnikova, M L Gershanovich, T Iu Semiglazova
ABSTRACT
In a retrospective study during the primary mode MOPP to primary patients LH II/IVAB stages with a poor prognosis rate of CR, 5--and 10-year DFS, OS was 69%, 71% and 68%, 74% and 64%, ABVD--76%, 78%, 83% and 68%, BEACOPP-baseline--73%, 97%, 85% and 82%, respectively. When the program ran BEACOPP-baseline it was revealed higher rates of DFS compared with ABVD and MOPP. Higher OS rates were observed in the primary patients treated with BEACOPP-baseline compared with MOPP (p = 0.04). In terms of DFS and OS MOPP regimen did not differ from ABVD. Program ABVD and BEACOPP-baseline had no differences in terms of OS. The frequency of primary refractory forms of LH did not depend on the conducted regimens of PCT. Significantly less recurrences occurred during the regimen of BEACOPP-baseline compared to MOPP and ABVD. BEACOPP-baseline was accompanied by a more pronounced reversible hematologic toxicity. Against the background of the program BEACOPP-baseline, neutropenia of III-IV degree was detected in 32%, ABVD--16%, MOPP--13%.
Product Description
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dacarbazine, antineoplastic purine analog
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Procarbazine hydrochloride, ≥98% (HPLC)