MilliporeSigma
  • Enzymatic saccharification coupling with polyester recovery from cotton-based waste textiles by phosphoric acid pretreatment.
Enzymatic saccharification coupling with polyester recovery from cotton-based waste textiles by phosphoric acid pretreatment.
Bioresource technology (2013-01-15)
Fei Shen, Wenxiong Xiao, Lili Lin, Gang Yang, Yanzong Zhang, Shihuai Deng
ABSTRACT
In order to recycle the cotton-based waste textiles, a novel process was designed for pretreating waste textiles with phosphoric acid to recover polyester and fermentable sugar. The effects of pretreatment conditions including, phosphoric acid concentration, pretreatment temperature, time, and ratio of textiles and phosphoric acid were thoroughly investigated. Results indicated the mentioned four factors had significant influences on sugar and polyester recovery. Almost complete polyester recovery was achieved by enhancing phosphoric acid concentration, temperature and pretreatment time or reducing the ratio of textiles and phosphoric acid. However, these behaviors decreased the sugar recovery seriously. 100% polyester recovery with a maximum sugar recovery of 79.2% was achieved at the optimized conditions (85% phosphoric acid, 50°C, 7h, and the ratio of 1:15). According to the technical and cost-benefit analysis, it was technically feasible and potentially profitable to recover polyester and sugar from waste textiles by phosphoric acid pretreatment.
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O