MilliporeSigma
  • Covalent bonding of zeolitic imidazolate framework-90 to functionalized silica fibers for solid-phase microextraction.
Covalent bonding of zeolitic imidazolate framework-90 to functionalized silica fibers for solid-phase microextraction.
Chemical communications (Cambridge, England) (2013-02-08)
Li-Qing Yu, Xiu-Ping Yan
ABSTRACT
Here we report a covalent bonding approach to fabricate a robust metal-organic framework ZIF-90 coating for solid-phase microextraction. The ZIF-90 bonded fiber exhibits high enhancement factors, wide linearity, excellent reproducibility, and good lifetime for the studied polar endocrine disruptors.
Product Description
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~1,200, 45 wt. % in H2O
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
4-Octylphenol, 99%
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
4-Octylphenol, analytical standard
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Supelco
Poly(acrylic acid sodium salt), analytical standard, for GPC, 16,000