MilliporeSigma
  • Diffusion-controlled detection of glucose with microelectrodes in mediated bioelectrocatalytic oxidation.
Diffusion-controlled detection of glucose with microelectrodes in mediated bioelectrocatalytic oxidation.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2013-03-12)
Tatsuo Noda, Mizue Wanibuchi, Yuki Kitazumi, Seiya Tsujimura, Osamu Shirai, Masahiro Yamamoto, Kenji Kano
ABSTRACT
This paper describes a diffusion-controlled electrolysis of glucose with mediated bioelectrocatalysis at microdisk-electrodes. Under conditions of an extremely fast enzyme reaction, compared with the diffusion of glucose, the current in potential-step chronoamperometry reaches an almost steady state within 10 s, and is proportional to the glucose concentration. The current can be detected at +0.1 V (vs. Ag|AgCl) with 9,10-phenanthrenequinone as a mediator. The addition of carboxymethylcellulose increased the linear range up to 10 mM.
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~90,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~700,000
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity