MilliporeSigma
  • Estrogen receptor α36 mediates a bone-sparing effect of 17β-estrodiol in postmenopausal women.
Estrogen receptor α36 mediates a bone-sparing effect of 17β-estrodiol in postmenopausal women.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2010-06-26)
Hui Xie, Mei Sun, Xiao-Bo Liao, Ling-Qing Yuan, Zhi-Feng Sheng, Ji-Cai Meng, Dan Wang, Zhi-Yong Yu, Lei-Yi Zhang, Hou-De Zhou, Xiang-Hang Luo, Hui Li, Xian-Ping Wu, Qi-You Wei, Si-Yuan Tang, Zhao-Yi Wang, Er-Yuan Liao
ABSTRACT
Recently, a membrane-based estrogen receptor (ER), ER-α36, was identified and cloned that transduces membrane-initiated estrogen signaling such as activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase (MAPK/ERK) signaling pathway. Here we show that the postmenopausal level of estradiol (E2) induces mitogenic, antiapoptotic, and antiosteogenic effects and proapoptotic effects in postmenopausal osteoblasts and osteoclasts with high levels of ER-α36 expression, respectively. We also found that ER-α36 mediated the effects of postmenopausal-level E(2) on proliferation, apoptosis, and differentiation of osteoblasts through transient activation of the MAPK/ERK pathway, whereas ER-α36-mediated postmenopausal-level E(2) induces apoptosis of osteoclasts through prolonged activation of the MAPK/ERK pathway with the involvement of reactive oxygen species. We also show that the levels of ER-α36 expression in bone are positively associated with bone mineral density but negatively associated with bone biochemical markers in postmenopausal women. Thus the higher levels of ER-α36 expression are required for preserving bone mass in postmenopausal and menopausal women who become osteoporotic if ER-α36-mediated activities are dysregulated.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%