MilliporeSigma
  • Functionalization of the tetrairon cluster Cp4Fe4(CO)4 with fulleropyrrolidine.
Functionalization of the tetrairon cluster Cp4Fe4(CO)4 with fulleropyrrolidine.
Dalton transactions (Cambridge, England : 2003) (2013-07-13)
Wen-Yann Yeh, Sung-Hua Wu
ABSTRACT
Condensation of the formyl group of Cp3Fe4(CO)4(C5H4CHO) with N-methylglycine and C60 produces Cp3Fe4(CO)4(C5H4-CHNMeCH2C60) (1), where one cyclopentadienyl ring of the tetrairon cluster is linked to a fulleropyrrolidinyl moiety. A similar reaction with the bisformyl cluster Cp2Fe4(CO)4(C5H4CHO)2 affords Cp2Fe4(CO)4(C5H4-CHNMeCH2C60)2 (2). Compounds 1 and 2 have been characterized by mass, IR, UV-Vis, and NMR spectroscopy. The electrochemistry of 1 and 2 was investigated by cyclic voltammetry and differential pulse voltammetry.
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Fullerene-C60, 99.5%
Sigma-Aldrich
Fullerene-C60, 98%
Sigma-Aldrich
Fullerene-C60, sublimed, 99.9%