MilliporeSigma
  • Surface modification of mammalian cells with stimuli-responsive polymers.
Surface modification of mammalian cells with stimuli-responsive polymers.
Chemical communications (Cambridge, England) (2013-07-26)
Yasuhiko Iwasaki, Mizuki Sakiyama, Shuetsu Fujii, Shin-ichi Yusa
ABSTRACT
In order to introduce alternative functions into mammalian cells and control them under ambient conditions, poly(N-isopropyl acrylamide) (PNIPAM) was immobilized on the cell surface. Cellular aggregation could be regulated by temperature change. In addition, separation of PNIPAM-conjugated glycoproteins was successfully performed.
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000