MilliporeSigma
  • Effectiveness of nonpharmacologic treatments for acute seasonal allergic conjunctivitis.
Effectiveness of nonpharmacologic treatments for acute seasonal allergic conjunctivitis.
Ophthalmology (2013-09-28)
Paramdeep S Bilkhu, James S Wolffsohn, Shehzad A Naroo, Louise Robertson, Roy Kennedy
ABSTRACT
To investigate whether artificial tears and cold compress alone or in combination provide a treatment benefit and whether they were as effective as or could enhance topical antiallergic medication. Randomized, masked clinical trial. Eighteen subjects (mean age, 29.5±11.0 years) allergic to grass pollen. Controlled exposure to grass pollen using an environmental chamber to stimulate an ocular allergic reaction followed by application of artificial tears (ATs), 5 minutes of cold compress (CC), ATs combined with CC, or no treatment applied at each separate visit in random order. A subset of 11 subjects also had epinastine hydrochloride (EH) applied alone and combined with CC in random order or instillation of a volume-matched saline control. Bulbar conjunctival hyperemia, ocular surface temperature, and ocular symptoms repeated before and every 10 minutes after treatment for 1 hour. Bulbar conjunctival hyperemia and ocular symptoms decreased and temperature recovered to baseline faster with nonpharmaceutical treatments compared with no treatment (P <0.05). Artificial tears combined with CC reduced hyperemia more than other treatments (P <0.05). The treatment effect of EH was enhanced by combining it with a CC (P <0.001). Cold compress combined with ATs or EH lowered the antigen-raised ocular surface temperature to less than the pre-exposure baseline. Artificial tear instillation alone or CC combined with ATs or EH significantly reduced the temperature (P <0.05). Cold compress combined with ATs or EH had a similar cooling effect (P >0.05). At all measurement intervals, symptoms were reduced for both EH and EH combined with CC than CC or ATs alone or in combination (P <0.014). After controlled exposure to grass pollen, CC and AT treatment showed a therapeutic effect on the signs and symptoms of allergic conjunctivitis. A CC enhanced the use of EH alone and was the only treatment to reduce symptoms to baseline within 1 hour of antigenic challenge. Signs of allergic conjunctivitis generally were reduced most by a combination of a CC in combination with ATs or EH.
Product Description
Epinastine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Epinastine hydrochloride, ≥98% (HPLC), solid