MilliporeSigma
Studies on the effect of antidiabetic drugs on collagen in rats.
Pakistan journal of biological sciences : PJBS (2014-02-12)
U M D Lekshmi, P N Reddy
ABSTRACT
To assess the effects of antidiabetic drugs on collagen, the study has been carried out. However, at present, no conclusive data available on the In vivo effect of antidiabetic drugs in collagen. It was therefore thought worthwhile to undertake a systemic study on the metabolic turnover of connective tissue proteins, particularly collagen which will help to understand the various changes taking place in conventional antidiabetic drug treated rats. The hydroxyproline content was determined by the method of Neumann and Logan. There was no significant difference in mean body weight of animals in treated groups when compared to control animals on 50th day. In drug treated group G5, urinary excretion of hydroxyproline was slightly increased in contrast to that of the rats in other groups and showed a significant difference (p < 0.001) between control and G5 group. The proportion of soluble collagen in treated group was somewhat higher than in the control but while taking the significance value there are no statistically significant changes. Liver showed the highest concentration of 0.98 g collagen/100 g tissue, whereas kidney and heart showed 0.57 and 0.72 g collagen/100 g tissue. Observations of the present study collectively indicate that the anabolic rate of collagen metabolism is not altered significantly by the administration of antidiabetic drugs in normal rats.
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Supelco
trans-4-Hydroxy-L-proline, analytical standard
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance