MilliporeSigma
  • High miR-21 expression from FFPE tissues is associated with poor survival and response to adjuvant chemotherapy in colon cancer.
High miR-21 expression from FFPE tissues is associated with poor survival and response to adjuvant chemotherapy in colon cancer.
International journal of cancer (2013-10-15)
Naohide Oue, Katsuhiro Anami, Aaron J Schetter, Markus Moehler, Hirokazu Okayama, Mohammed A Khan, Elise D Bowman, Annett Mueller, Arno Schad, Manabu Shimomura, Takao Hinoi, Kazuhiko Aoyagi, Hiroki Sasaki, Masazumi Okajima, Hideki Ohdan, Peter R Galle, Wataru Yasui, Curtis C Harris
ABSTRACT
Colon cancer (CC) is a leading cause of cancer mortality. Novel biomarkers are needed to identify CC patients at high risk of recurrence and those who may benefit from therapeutic intervention. The aim of this study is to investigate if miR-21 expression from RNA isolated from formalin-fixed paraffin-embedded (FFPE) tissue sections is associated with prognosis and therapeutic outcome for patients with CC. The expression of miR-21 was measured by quantitative reverse transcriptase-polymerase chain reaction in a Japanese cohort (stage I-IV, n = 156) and a German cohort (stage II, n = 145). High miR-21 expression in tumors was associated with poor survival in both the stage II/III Japanese (p = 0.0008) and stage II German (p = 0.047) cohorts. These associations were independent of other clinical covariates in multivariable models. Receipt of adjuvant chemotherapy was not beneficial in patients with high miR-21 in either cohort. In the Japanese cohort, high miR-21 expression was significantly associated with poor therapeutic outcome (p = 0.0001) and adjuvant therapy was associated with improved survival in patients with low miR-21 (p = 0.001). These results suggest that miR-21 is a promising biomarker to identify patients with poor prognosis and can be accurately measured in FFPE tissues. The expression of miR-21 may also identify patients who will benefit from adjuvant chemotherapy.
Sigma-Aldrich
Formaldehyde neutral buffer solution, 3.7% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde neutral buffer solution, 7.5% formaldehyde in H2O
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)