MilliporeSigma
  • Lack of identifiable biologic behavior in a series of porcine mesh explants.
Lack of identifiable biologic behavior in a series of porcine mesh explants.
Surgery (2014-05-27)
Gayan S De Silva, David M Krpata, Yue Gao, Cory N Criss, James M Anderson, Hooman T Soltanian, Michael J Rosen, Yuri W Novitsky
ABSTRACT
Biologic matrices used in abdominal wall reconstruction are purported to undergo remodeling into connective tissue resembling native collagen. Key steps in that process include inflammatory response at the mesh/tissue interface, cellular penetration, and neovascularization of the matrix, followed by fibroblast proliferation and collagen deposition. We aimed to examine the concept of biologic mesh remodeling/regeneration in a series of explanted porcine biologic meshes. A cohort of patients who underwent removal of porcine biologic mesh was identified in a prospective database. Mesh/tissue samples were analyzed using standard hematoxylin/eosin and Masson's trichrome staining. Main outcome measures included: inflammatory response at the mesh/tissue interface, foreign body reaction (FBR), cellular penetration, neovascularization, and new collagen deposition. All evaluations were performed by a blinded senior pathologist using established grading scales. A total of 14 cases with implant time ranging from 4 to 33 months were identified and analyzed. All meshes were placed as intraperitoneal underlay. There were 7 non-cross-linked and 7 cross-linked grafts. Cross-linked grafts were associated with mild FBR and moderate fibrous capsule formation. Similarly, non-cross-linked grafts had mild-to-moderate FBR and encapsulation. Furthermore, non-cross-linked grafts were associated with no neovascularization and minimal peripheral mesh neocellularization. Cross-linked grafts demonstrated neither neovascularization nor neocellularization. Although no grafts were associated with any quantifiable new collagen deposition within the porcine biologic matrix, minimal biodegradation/remodeling was observed at the periphery of the non-cross-linked grafts only. The biologic behavior of porcine meshes is predicated on their ability to undergo mesh remodeling with resorption and new collagen deposition. In the largest series of human biologic explants, we detected no evidence of xenograft remodeling, especially in the cross-linked group. Although underlay mesh placement and other patient factors may have contributed to our findings, the concept of porcine biologic mesh regeneration does not seem to be prevalent in the clinical setting.
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture