MilliporeSigma
  • Antigen-specific immune responses and clinical outcome after vaccination with glioma-associated antigen peptides and polyinosinic-polycytidylic acid stabilized by lysine and carboxymethylcellulose in children with newly diagnosed malignant brainstem and nonbrainstem gliomas.
Antigen-specific immune responses and clinical outcome after vaccination with glioma-associated antigen peptides and polyinosinic-polycytidylic acid stabilized by lysine and carboxymethylcellulose in children with newly diagnosed malignant brainstem and nonbrainstem gliomas.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2014-06-04)
Ian F Pollack, Regina I Jakacki, Lisa H Butterfield, Ronald L Hamilton, Ashok Panigrahy, Douglas M Potter, Angela K Connelly, Sharon A Dibridge, Theresa L Whiteside, Hideho Okada
ABSTRACT
Diffuse brainstem gliomas (BSGs) and other high-grade gliomas (HGGs) of childhood carry a dismal prognosis despite current treatments, and new therapies are needed. Having identified a series of glioma-associated antigens (GAAs) commonly overexpressed in pediatric gliomas, we initiated a pilot study of subcutaneous vaccinations with GAA epitope peptides in HLA-A2-positive children with newly diagnosed BSG and HGG. GAAs were EphA2, interleukin-13 receptor alpha 2 (IL-13Rα2), and survivin, and their peptide epitopes were emulsified in Montanide-ISA-51 and given every 3 weeks with intramuscular polyinosinic-polycytidylic acid stabilized by lysine and carboxymethylcellulose for eight courses, followed by booster vaccinations every 6 weeks. Primary end points were safety and T-cell responses against vaccine-targeted GAA epitopes. Treatment response was evaluated clinically and by magnetic resonance imaging. Twenty-six children were enrolled, 14 with newly diagnosed BSG treated with irradiation and 12 with newly diagnosed BSG or HGG treated with irradiation and concurrent chemotherapy. No dose-limiting non-CNS toxicity was encountered. Five children had symptomatic pseudoprogression, which responded to dexamethasone and was associated with prolonged survival. Only two patients had progressive disease during the first two vaccine courses; 19 had stable disease, two had partial responses, one had a minor response, and two had prolonged disease-free status after surgery. Enzyme-linked immunosorbent spot analysis in 21 children showed positive anti-GAA immune responses in 13: to IL-13Rα2 in 10, EphA2 in 11, and survivin in three. GAA peptide vaccination in children with gliomas is generally well tolerated and has preliminary evidence of immunologic and clinical responses. Careful monitoring and management of pseudoprogression is essential.
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~90,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Supelco
L-Lysine, analytical standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Lysine acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~700,000
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material