MilliporeSigma
  • A new X-ray activated nanoparticle photosensitizer for cancer treatment.
A new X-ray activated nanoparticle photosensitizer for cancer treatment.
Journal of biomedical nanotechnology (2014-07-16)
Lun Ma, Xiaoju Zou, Wei Chen
ABSTRACT
Photodynamic therapy (PDT) has attracted ever-growing attention as a promising modality in the treatment of cancer. However, due to the poor tissue penetration by light, PDT has rarely been applied for treating deep-seated tumors. This problem can be solved if photosensitizers are activated by X-rays, which are able to penetrate deeply into tissues. Previous attempts using X-rays to activate photosensitizers were not very successful, since the traditional PDT photosensitizers could not efficiently be activated with X-rays. Here we exploit copper-cysteamine complex (Cu-Cy) nanoparticles as a new type of photosensitizer activated by X-ray for cancer treatment. The newly invented Cu-Cy nanoparticles can be activated directly by X-rays to produce singlet oxygen. In vitro and in vivo study on human breast cancer cells (MCF-7) have shown significant cell destruction using Cu-Cy nanoparticles under X-ray activation. The Cu-Cy nanoparticles are a novel and potent X-ray activated photosensitizer which can enable PDT for deep cancer treatment.
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Continuous cast copper (O), BCR®, certified reference material, rod
Copper, tube, 200mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, tube, 500mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Copper, tube, 500mm, outside diameter 0.65mm, inside diameter 0.21mm, wall thickness 0.22mm, as drawn, 99.9%
Copper, tube, 200mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 3.5mm, inside diameter 1.56mm, wall thickness 0.97mm, as drawn, 99.9%
Copper, tube, 500mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 20m coil, thickness 0.010mm, coil width 25mm, hard, 99.95+%
Copper, tube, 200mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.9%
Copper, tube, 500mm, outside diameter 0.25mm, inside diameter 0.05mm, wall thickness 0.1mm, as drawn, 99.9%
Copper, tube, 500mm, outside diameter 10.4mm, inside diameter 10mm, wall thickness 0.2mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.7mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.075mm, half hard, 99.95+%
Copper, tube, 200mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 5m coil, thickness 0.1mm, coil width 100mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10mm disks, thickness 1.0mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 15mm disks, thickness 0.25mm, annealed, 99.95+%