MilliporeSigma
  • Forkhead Box F1 represses cell growth and inhibits COL1 and ARPC2 expression in lung fibroblasts in vitro.
Forkhead Box F1 represses cell growth and inhibits COL1 and ARPC2 expression in lung fibroblasts in vitro.
American journal of physiology. Lung cellular and molecular physiology (2014-09-28)
Sara Melboucy-Belkhir, Pauline Pradère, Sara Tadbiri, Stéfanie Habib, Antoine Bacrot, Stéphanie Brayer, Bernard Mari, Valérie Besnard, Arnaud Mailleux, Andreas Guenther, Yves Castier, Hervé Mal, Bruno Crestani, Laurent Plantier
ABSTRACT
Aberrant expression of master phenotype regulators or alterations in their downstream pathways in lung fibroblasts may play a central role in idiopathic pulmonary fibrosis (IPF). Interrogating IPF fibroblast transcriptome datasets, we identified Forkhead Box F1 (FOXF1), a DNA-binding protein required for lung development, as a candidate actor in IPF. Thus we determined FOXF1 expression levels in fibroblasts cultured from normal or IPF lungs in vitro, and explored FOXF1 functions in these cells using transient and stable loss-of-function and gain-of-function models. FOXF1 mRNA and protein were expressed at higher levels in IPF fibroblasts compared with normal fibroblasts (mRNA: +44%, protein: +77%). Immunohistochemistry showed FOXF1 expression in nuclei of bronchial smooth muscle cells, endothelial cells, and lung fibroblasts including fibroblastic foci of IPF lungs. In normal lung fibroblasts, FOXF1 repressed cell growth and expression of collagen-1 (COL1) and actin-related protein 2/3 complex, subunit 2 (ARPC2). ARPC2 knockdown inhibited cell growth and COL1 expression, consistent with FOXF1 acting in part through ARPC2 repression. In IPF fibroblasts, COL1 and ARPC2 repression by FOXF1 was blunted, and FOXF1 did not repress growth. FOXF1 expression was induced by the antifibrotic mediator prostaglandin E2 and repressed by the profibrotic cytokine transforming growth factor-β1 in both normal and IPF lung fibroblasts. Ex vivo, FOXF1 knockdown conferred CCL-210 lung fibroblasts the ability to implant in uninjured mouse lungs. In conclusion, FOXF1 functions and regulation were consistent with participation in antifibrotic pathways. Alterations of pathways downstream of FOXF1 may participate to fibrogenesis in IPF fibroblasts.
Product Description
Supelco
Staurosporine, VETRANAL®, analytical standard
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Supelco
Cycloheximide, PESTANAL®, analytical standard
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Fas Antibody (human, activating), clone CH11, clone CH11, Upstate®, from mouse
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission