MilliporeSigma
  • Regulation of auxin, abscisic acid and salicylic acid levels by ascorbate application under heat stress in sensitive and tolerant maize leaves.
Regulation of auxin, abscisic acid and salicylic acid levels by ascorbate application under heat stress in sensitive and tolerant maize leaves.
Acta biologica Hungarica (2014-12-06)
Burcu Seckin Dinler, Emel Demir, Yasemin Ozdener Kompe
ABSTRACT
In the present study, the effect of ascorbic acid (5 mM) on some physiological parameters and three hormones (auxin, abscisic acid, salicylic acid) was determined under heat stress (40 °C) in maize tolerant cv. (MAY 69) and sensitive cv. SHEMAL (SH) at 0 h, 4 h and 8 h. Heat stress reduced total chlorophyll content (CHL), relative water content (RWC) and stomatal conductance (gs) in SH but did not lead to changes in MAY 69 at 4 h and 8 h. However, pretreatment with ascorbic acid increased (CHL), (RWC) and (gs) in SH under heat stress while it reduced MDA content significantly in both cv. We also observed that heat stress led to a reduction in SA level but increased ABA and IAA levels in SH, whereas it increased SA and IAA levels but did not change ABA level in MAY 69 at 4 h. Furthermore, in SH, ASC application under heat stress increased SA level and decreased IAA and ABA levels at 4 h, but it had no effect on SA and ABA at 8 h.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Water, ACS reagent, for ultratrace analysis
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent