MilliporeSigma
  • Isolation of neural progenitor cells from the human adult subventricular zone based on expression of the cell surface marker CD271.
Isolation of neural progenitor cells from the human adult subventricular zone based on expression of the cell surface marker CD271.
Stem cells translational medicine (2014-03-08)
Miriam E van Strien, Jacqueline A Sluijs, Brent A Reynolds, Dennis A Steindler, Eleonora Aronica, Elly M Hol
ABSTRACT
Neural progenitor cells (NPCs) in the subventricular zone (SVZ) hold promise for future therapy for neurodegenerative disorders, because the stimulation of adult neurogenesis could potentially restore the function of degenerating neurons and glia. To obtain more knowledge on these NPCs, we developed a method to specifically isolate NPCs from postmortem adult human brains based on the expression of the specific human adult neural stem/progenitor cell marker glial fibrillary acidic protein δ (GFAPδ). An extensive immunophenotyping analysis for cell surface markers resulted in the observation that CD271 was limited to the SVZ-derived GFAPδ-positive cells. CD271(+) cells developed into neurospheres and could be differentiated into astrocytes, neurons, and oligodendrocytes. We are the first to show that a pure population of NPCs can be isolated from the adult human SVZ, which is highly instrumental for developing future therapies based on stimulating endogenous SVZ neurogenesis.