MilliporeSigma
  • The phenotypic and genotypic spectra of ichthyosis with confetti plus novel genetic variation in the 3' end of KRT10: from disease to a syndrome.
The phenotypic and genotypic spectra of ichthyosis with confetti plus novel genetic variation in the 3' end of KRT10: from disease to a syndrome.
JAMA dermatology (2014-09-12)
Iris Spoerri, Michela Brena, Julie De Mesmaeker, Nina Schlipf, Judith Fischer, Gianluca Tadini, Peter H Itin, Bettina Burger
ABSTRACT
Ichthyosis with confetti (IWC) is a genodermatosis caused by dominant negative mutations in the gene encoding keratin 10 (KRT10). We investigated clinical and genetic details of a substantial number of patients with IWC in order to define major and minor criteria for diagnosis of this rare disorder. Parallel clinical investigation of 6 patients with IWC revealed a novel spectrum of phenotypes. We found several features that qualify as major criteria for diagnosis, which are clearly and consistently associated with the condition. These included malformation of ears, hypoplasia of mammillae, and dorsal acral hypertrichosis. Genetic analysis of patients revealed several different frameshift mutations in intron 6 or exon 7 of KRT10. Analysis of this locus in 17 unrelated control individuals revealed 2 novel polymorphisms of KRT10. We present for the first time to our knowledge the spectrum of clinical variability of IWC in 6 patients with confirmed mutations in KRT10. From this, we have extracted major and minor criteria to aid early and correct clinical diagnosis. Ectodermal malformations, present in all patients, suggest a novel classification of IWC as a syndrome. There is remarkable genetic variation at the IWC disease locus within control individuals from the general population.