MilliporeSigma
Cbfb regulates bone development by stabilizing Runx family proteins.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2014-09-30)
Xin Qin, Qing Jiang, Yuki Matsuo, Tetsuya Kawane, Hisato Komori, Takeshi Moriishi, Ichiro Taniuchi, Kosei Ito, Yosuke Kawai, Satoshi Rokutanda, Shinichi Izumi, Toshihisa Komori
ABSTRACT
Runx family proteins, Runx1, Runx2, and Runx3, play important roles in skeletal development. Runx2 is required for osteoblast differentiation and chondrocyte maturation, and haplodeficiency of RUNX2 causes cleidocranial dysplasia, which is characterized by open fontanelles and sutures and hypoplastic clavicles. Cbfb forms a heterodimer with Runx family proteins and enhances their DNA-binding capacity. Cbfb-deficient (Cbfb(-/-) ) mice die at midgestation because of the lack of fetal liver hematopoiesis. We previously reported that the partial rescue of hematopoiesis in Cbfb(-/-) mice revealed the requirement of Cbfb in skeletal development. However, the precise functions of Cbfb in skeletal development still remain to be clarified. We deleted Cbfb in mesenchymal cells giving rise to both chondrocyte and osteoblast lineages by mating Cbfb(fl/fl) mice with Dermo1 Cre knock-in mice. Cbfb(fl/fl/Cre) mice showed dwarfism, both intramembranous and endochondral ossifications were retarded, and chondrocyte maturation and proliferation and osteoblast differentiation were inhibited. The differentiation of chondrocytes and osteoblasts were severely inhibited in vitro, and the reporter activities of Ihh, Col10a1, and Bglap2 promoter constructs were reduced in Cbfb(fl/fl/Cre) chondrocytes or osteoblasts. The proteins of Runx1, Runx2, and Runx3 were reduced in the cartilaginous limb skeletons and calvariae of Cbfb(fl/fl/Cre) embryos compared with the respective protein in the respective tissue of Cbfb(fl/fl) embryos at E15.5, although the reduction of Runx2 protein in calvariae was much milder than that in cartilaginous limb skeletons. All of the Runx family proteins were severely reduced in Cbfb(fl/fl/Cre) primary osteoblasts, and Runx2 protein was less stable in Cbfb(fl/fl/Cre) osteoblasts than Cbfb(fl/fl) osteoblasts. These findings indicate that Cbfb is required for skeletal development by regulating chondrocyte differentiation and proliferation and osteoblast differentiation; that Cbfb plays an important role in the stabilization of Runx family proteins; and that Runx2 protein stability is less dependent on Cbfb in calvariae than in cartilaginous limb skeletons.
Product Description
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Supelco
Cycloheximide, PESTANAL®, analytical standard
Roche
X-tremeGENE 9 DNA Transfection Reagent, Polymer reagent for transfecting common cell lines