MilliporeSigma
  • Formulation and characterization of atropine sulfate in albumin-chitosan microparticles for in vivo ocular drug delivery.
Formulation and characterization of atropine sulfate in albumin-chitosan microparticles for in vivo ocular drug delivery.
Journal of pharmaceutical sciences (2015-02-06)
Richard T Addo, Kwame G Yeboah, Rodney C Siwale, Aladin Siddig, Alphia Jones, Ruhi V Ubale, Janet Akande, Henry Nettey, Neil J Patel, Evelyn Addo, Martin J D'Souza
ABSTRACT
The overall study goal was to produce a microparticle formulation containing atropine sulfate for ocular administration with improved efficacy and lower side effects, compared with that of the standard marketed atropine solution. The objective was to prepare an atropine sulfate-loaded bovine serum albumin-chitosan microparticle that would have longer contact time on the eyes as well as better mydriatic and cycloplegic effect using a rabbit model. The microparticle formulation was prepared by method of spray-drying technique. The percent drug loading and encapsulation efficiency were assessed using a USP (I) dissolution apparatus. The particle sizes and zeta potential were determined using laser scattering technique and the surface morphology of the microparticles was determined using a scanning electron microscope. The product yield was calculated from relative amount of material used. In vitro cytotoxicity and uptake by human corneal epithelial cells were examined using AlamarBlue and confocal microscopy. The effects of the microparticle formulation on mydriasis in comparison with the marketed atropine sulfate solution were evaluated in rabbit eyes. The prepared microparticle formulation had ideal physicochemical characteristics for delivery into the eyes. The in vivo studies showed that the microparticles had superior effects on mydriasis in rabbits than the marketed solutions
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Supelco
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O