MilliporeSigma
  • LYST affects lysosome size and quantity, but not trafficking or degradation through autophagy or endocytosis.
LYST affects lysosome size and quantity, but not trafficking or degradation through autophagy or endocytosis.
Traffic (Copenhagen, Denmark) (2014-09-13)
Petter Holland, Maria L Torgersen, Kirsten Sandvig, Anne Simonsen
ABSTRACT
Mutations in the large BEACH domain-containing protein LYST causes Chediak-Higashi syndrome. The diagnostic hallmark is enlarged lysosomes and lysosome-related organelles in various cell types. Dysfunctional secretion of enlarged lysosome-related organelles has been observed in cells with mutations in LYST, but the capacity of the enlarged lysosomes to degrade endogenous proteins has not been studied. Here, we show for the first time that small interfering RNA-depletion of LYST in human cell lines recapitulates the LYST mutant phenotype of enlarged lysosomes. We found no evidence for an effect of LYST depletion on autophagy or endocytic degradation. Autophagosomes are formed in normal size and quantities and are able to fuse to the enlarged lysosomes, leading to normal rates of degradation. Degradation of the epidermal growth factor receptor (EGFR) was similarly not affected, indicating that the enlarged lysosomes are fully functional in degrading endogenous proteins. Retrograde trafficking of toxins as well as the localization of transporters of lysosomal proteins, adaptor protein-3 (AP-3) and cation-independent mannose-6-phosphate receptor (CI-MPR), were all found to be unaffected by LYST. Quantitative analysis of the enlarged lysosomes shows that LYST depletion causes a reduction in vesicle quantity per cell, while the total enzymatic content and vesicular pH are unaffected, supporting a role for LYST in lysosomal fission and/or fusion events.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Glycine
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O