MilliporeSigma
  • The neuroendocrine phenotype of gastric myofibroblasts and its loss with cancer progression.
The neuroendocrine phenotype of gastric myofibroblasts and its loss with cancer progression.
Carcinogenesis (2014-04-09)
Silvia Balabanova, Chris Holmberg, Islay Steele, Bahram Ebrahimi, Lucille Rainbow, Ted Burdyga, Cathy McCaig, Lazso Tiszlavicz, Nantaporn Lertkowit, Olivier T Giger, Simon Oliver, Ian Prior, Rod Dimaline, Deborah Simpson, Rob Beynon, Peter Hegyi, Timothy C Wang, Graham J Dockray, Andrea Varro
ABSTRACT
Stromal cells influence cancer progression. Myofibroblasts are an important stromal cell type, which influence the tumour microenvironment by release of extracellular matrix (ECM) proteins, proteases, cytokines and chemokines. The mechanisms of secretion are poorly understood. Here, we describe the secretion of marker proteins in gastric cancer and control myofibroblasts in response to insulin-like growth factor (IGF) stimulation and, using functional genomic approaches, we identify proteins influencing the secretory response. IGF rapidly increased myofibroblast secretion of an ECM protein, TGFβig-h3. The secretory response was not blocked by inhibition of protein synthesis and was partially mediated by increased intracellular calcium (Ca(2+)). The capacity for evoked secretion was associated with the presence of dense-core secretory vesicles and was lost in cells from patients with advanced gastric cancer. In cells responding to IGF-II, the expression of neuroendocrine marker proteins, including secretogranin-II and proenkephalin, was identified by gene array and LC-MS/MS respectively, and verified experimentally. The expression of proenkephalin was decreased in cancers from patients with advanced disease. Inhibition of secretogranin-II expression decreased the secretory response to IGF, and its over-expression recovered the secretory response consistent with a role in secretory vesicle biogenesis. We conclude that normal and some gastric cancer myofibroblasts have a neuroendocrine-like phenotype characterized by Ca(2+)-dependent regulated secretion, dense-core secretory vesicles and expression of neuroendocrine marker proteins; loss of the phenotype is associated with advanced cancer. A failure to regulate myofibroblast protein secretion may contribute to cancer progression.
Product Description
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%