MilliporeSigma
Hormonal induction and roles of Disabled-2 in lactation and involution.
PloS one (2014-11-02)
Wensi Tao, Robert Moore, Elizabeth R Smith, Xiang-Xi Xu
ABSTRACT
Disabled-2 (Dab2) is a widely expressed endocytic adaptor that was first isolated as a 96 KDa phospho-protein, p96, involved in MAPK signal transduction. Dab2 expression is lost in several cancer types including breast cancer, and Dab2 is thought to have a tumor suppressor function. In mammary epithelia, Dab2 was induced upon pregnancy and further elevated during lactation. We constructed mutant mice with a mosaic Dab2 gene deletion to bypass early embryonic lethality and to investigate the roles of Dab2 in mammary physiology. Loss of Dab2 had subtle effects on lactation, but Dab2-deficient mammary glands showed a strikingly delayed cell clearance during involution. In primary cultures of mouse mammary epithelial cells, Dab2 proteins were also induced by estrogen, progesterone, and/or prolactin. Dab2 null mammary epithelial cells were refractory to growth suppression induced by TGF-beta. However, Dab2 deletion did not affect Smad2 phosphorylation; rather TGF-beta-stimulated MAPK activation was enhanced in Dab2-deficient cells. We conclude that Dab2 expression is induced by hormones and Dab2 plays a role in modulating TGF-beta signaling to enhance apoptotic clearance of mammary epithelial cells during involution.
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Progesterone, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Oxytocin, European Pharmacopoeia (EP) Reference Standard
USP
Oxytocin, United States Pharmacopeia (USP) Reference Standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Supelco
Progesterone, VETRANAL®, analytical standard