MilliporeSigma
  • Outbreak of Microsporum audouinii in Munich--the return of infectious fungi in Germany.
Outbreak of Microsporum audouinii in Munich--the return of infectious fungi in Germany.
Mycoses (2014-09-02)
Alexander Zink, Vasileios Papanagiotou, Antonia Todorova, Hans-Peter Seidl, Andrea Niedermeier, Johannes Ring, Claudia Traidl-Hoffmann
ABSTRACT
After experiencing an unusually high number of Microsporum (M.) audouinii infections at our hospital within only a few weeks, we began to investigate and control an outbreak in Munich, Germany. Main goals of our health management were to treat infected persons, identify extent and form of transmission and to prevent new infections. We analysed data from structured interviews with patients and mycological cultures of swabs taken of patients and investigated involved public facilities. Outbreak management included antifungal treatment of patients, decontamination of affected facilities, the introduction of a temporary kindergarten ban for M. audouinii positive children and the organisation of educational meetings. Between March and August 2011, 16 children and 4 adults were identified with M. audouinii infections. The fungus was brought to Munich by the index patients from a family vacation in Africa and then spread to fellow children in kindergarten and subsequently to their families. All patients were treated successfully and the epidemic was declared ceased after 40 weeks but causing considerable financial damage. Due to travelling and migration, M. audouinii infections will rise in Germany and Europe. Sufficient and sustainable strategies are needed for the management of future outbreaks of highly contagious fungi.
Product Description
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Chloramphenicol, γ-irradiated
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard