MilliporeSigma
  • Amphiphilic polyvinyl alcohol adsorbent for the removal of low-density lipoprotein.
Amphiphilic polyvinyl alcohol adsorbent for the removal of low-density lipoprotein.
Artificial cells, nanomedicine, and biotechnology (2014-05-13)
Yao Ting Yu, Huijun Zhu, Shenqi Wang
ABSTRACT
Spacer can effectively reduce the steric hindrance and synergistic effect of the hydrophilic and hydrophobic ligands immobilized in adsorbents can improve the specific adsorption for low-density lipoprotein (LDL). In this paper, in order to improve the adsorption capacity for the Low-density lipoprotein-cholesterol (LDL-C), specifically, amphiphilic adsorbent based on polyvinyl alcohol (PVA) containing cholesterol ligand and sulfonic dextran ligands was synthesized. All kinds of factors affecting the synthesis yield and adsorption properties were studied in detail. Results showed that the amphiphilic PVA adsorbent has higher adsorption capacity for total cholesterol (TC), (LDL-C), triglyceride (TG), and lower adsorption capacity, and percentage for high-density lipoprotein-cholesterol (HDL-C), while the ligand ratio of cholesterol to sulfonic ligands is 1.57:1, the adsorption percentage and adsorption capacity for TC, LDL-C, TG, and HDL-C were 54.4%, 67.6%, 42.5%, 10.4% and 4.02, 3.612, 2.154, 0.168 mg/g, respectively.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
(±)-Epichlorohydrin, analytical standard
Sigma-Aldrich
(±)-Epichlorohydrin, purum, ≥99% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M