MilliporeSigma
  • Asymmetric dimethylarginine triggers macrophage apoptosis via the endoplasmic reticulum stress pathway.
Asymmetric dimethylarginine triggers macrophage apoptosis via the endoplasmic reticulum stress pathway.
Molecular and cellular biochemistry (2014-09-12)
Dan Hong, Hai-Chao Gao, Xiang Wang, Ling-Fang Li, Chuan-Chang Li, Ying Luo, Kang-Kai Wang, Yong-Ping Bai, Guo-Gang Zhang
ABSTRACT
Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of nitric oxide synthase (NOS), is emerging as a key contributing factor in atherogenesis, a process in turn known to involve macrophage apoptosis. The aim of this study was to determine the effect of ADMA on macrophage apoptosis, with specific reference to the endoplasmic reticulum (ER) stress pathway. Macrophage apoptosis was evaluated by Annexin V- Propidium iodide (PI) and Hoechst 33258 staining assays. Levels of the ER stress marker glucose regulated protein 78 (GRP78) were characterized by western blot. Levels of the proapoptotic C/EBP-homologous protein (CHOP) were evaluated by western blot and reverse transcription polymerase chain reaction (RT-PCR), and caspase-4 activity was measured using a colorimetric protease assay kit. We observed ADMA dose- and time-dependent increases in macrophage levels of GRP78. Similar ADMA dose- and time-dependent increases were detected in intracellular caspase-4 activity and macrophage apoptosis, all of which were sensitive to treatment with siRNAs for protein kinase RNA-like ER kinase and inositol-requiring protein-1 (IRE1), the ADMA antagonist L-arginine, as well as inhibitors of eukaryotic translation initiation factor-2 (salubrinal), IRE1 (irestatin 9389), and c-Jun N-terminal kinase (SP600125). Our results indicate that ADMA triggers macrophage apoptosis via the ER stress pathway.
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)