MilliporeSigma
  • Determining trace amounts and the origin of formaldehyde impurity in Neisseria meningitidis A/C/Y/W-135-DT conjugate vaccine formulated in isotonic aqueous 1× PBS by improved C18-UPLC method.
Determining trace amounts and the origin of formaldehyde impurity in Neisseria meningitidis A/C/Y/W-135-DT conjugate vaccine formulated in isotonic aqueous 1× PBS by improved C18-UPLC method.
Journal of pharmaceutical and biomedical analysis (2015-02-11)
Seshu K Gudlavalleti, Erika N Crawford, Nhi N Tran, Dana J Orten, Jeffery D Harder, Jeeri R Reddy
ABSTRACT
The ability to accurately measure and report trace amounts of residual formaldehyde impurity in a vaccine product is not only critical in the product release but also a regulatory requirement. In many bacterial or viral vaccine manufacturing procedures, formaldehyde is used either at a live culture inactivation step or at a protein de-toxification step or at both. Reported here is a validated and improved C18-UPLC method (developed based on previously published C-8 HPLC method) to determine the traces of formaldehyde process impurity in a liquid form Neisseria meningitidis A/C/Y/W-135-DT conjugate vaccine formulated in isotonic aqueous 1× PBS. UPLC C-18 column and the conditions described distinctly resolved the 2,4-DNPH-HCHO adduct from the un-reacted 2,4-DNPH as detected by TUV detector at 360 nm. This method was shown to be compatible with PBS formulation and extremely sensitive (with a quantitation limit of 0.05 ppm) and aided to determine formaldehyde contamination sources by evaluating the in-process materials as a track-down analysis. Final nanogram levels of formaldehyde in the formulated single dose vialed vaccine mainly originated from the diphtheria toxoid carrier protein used in the production of the conjugate vaccine, whereas relative contribution from polysaccharide API was minimal.
Sigma-Aldrich
1-Decanethiol, 96%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
1-Decanethiol, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O