MilliporeSigma
  • Fc gamma receptor-TLR cross-talk elicits pro-inflammatory cytokine production by human M2 macrophages.
Fc gamma receptor-TLR cross-talk elicits pro-inflammatory cytokine production by human M2 macrophages.
Nature communications (2014-11-14)
Lisa T C Vogelpoel, Ivo S Hansen, Theo Rispens, Femke J M Muller, Toni M M van Capel, Maureen C Turina, Joost B Vos, Dominique L P Baeten, Martien L Kapsenberg, Esther C de Jong, Jeroen den Dunnen
ABSTRACT
M2 macrophages suppress inflammation in numerous disorders, including tumour formation, infection and obesity. However, the exact role of M2 macrophages in the context of several other diseases is still largely undefined. We here show that human M2 macrophages promote inflammation instead of suppressing inflammation on simultaneous exposure to complexed IgG (c-IgG) and TLR ligands, as occurs in the context of diseases such as rheumatoid arthritis (RA). c-IgG-TLR ligand co-stimulation of M2 macrophages selectively amplifies production of pro-inflammatory cytokines TNF-α, IL-1β and IL-6 and promotes Th17 responses, which all play a critical role in RA pathology. Induction of pro-inflammatory cytokines on c-IgG co-stimulation mainly depends on Fc gamma receptor IIa (FcγRIIa), which selectively amplifies cytokine gene transcription and induces caspase-1 activation. These data indicate that FcγR-TLR cross-talk may be targeted for treatment to attenuate inflammation in RA, by restoring the anti-inflammatory function of M2 macrophages.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium azide, BioXtra
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard