MilliporeSigma
  • Extracellular matrix protein 1 regulates cell proliferation and trastuzumab resistance through activation of epidermal growth factor signaling.
Extracellular matrix protein 1 regulates cell proliferation and trastuzumab resistance through activation of epidermal growth factor signaling.
Breast cancer research : BCR (2014-12-17)
Kyung-min Lee, Keesoo Nam, Sunhwa Oh, Juyeon Lim, Young-Pil Kim, Jong Won Lee, Jong-Han Yu, Sei-Hyun Ahn, Sung-Bae Kim, Dong-Young Noh, Taehoon Lee, Incheol Shin
ABSTRACT
Extracellular matrix protein 1 (ECM1) is a secreted glycoprotein with putative functions in cell proliferation, angiogenesis and differentiation. Expression of ECM1 in several types of carcinoma suggests that it may promote tumor development. In this study, we investigated the role of ECM1 in oncogenic cell signaling in breast cancer, and potential mechanisms for its effects. In order to find out the functional role of ECM1, we used the recombinant human ECM1 and viral transduction systems which stably regulated the expression level of ECM1. We examined the effect of ECM1 on cell proliferation and cell signaling in vitro and in vivo. Moreover, tissues and sera of patients with breast cancer were used to confirm the effect of ECM1. ECM1 protein was increased in trastuzumab-resistant (TR) cells, in association with trastuzumab resistance and cell proliferation. Through physical interaction with epidermal growth factor receptor (EGFR), ECM1 potentiated the phosphorylation of EGFR and extracellular signal-regulated kinase upon EGF treatment. Moreover, ECM1-induced galectin-3 cleavage through upregulation of matrix metalloproteinase 9 not only improved mucin 1 expression, but also increased EGFR and human epidermal growth factor receptor 3 protein stability as a secondary signaling. ECM1 has important roles in both cancer development and trastuzumab resistance in breast cancer through activation of EGFR signaling.
Product Description
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
2-Butanone, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Butanone, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material