MilliporeSigma
  • An integrated strategy based on UPLC-DAD-QTOF-MS for metabolism and pharmacokinetic studies of herbal medicines: Tibetan "Snow Lotus" herb (Saussurea laniceps), a case study.
An integrated strategy based on UPLC-DAD-QTOF-MS for metabolism and pharmacokinetic studies of herbal medicines: Tibetan "Snow Lotus" herb (Saussurea laniceps), a case study.
Journal of ethnopharmacology (2014-03-26)
Tao Yi, Lin Zhu, Yi-Na Tang, Jian-Ye Zhang, Zhi-Tao Liang, Jun Xu, Zhong-Zhen Zhao, Zhi-Ling Yu, Zhao-Xiang Bian, Zhi-Jun Yang, Hu-Biao Chen
ABSTRACT
Saussurea laniceps Hand.-Mazz. (SL) has long been used under the herbal name Tibetan "Snow Lotus" for the treatment of rheumatoid arthritis, stomachache and dysmenorrhea in Tibetan folk medicine. Since herbal medicine (HM) is a synergistical system with multiple components, both of the metabolism and pharmacokinetic studies of HM are interdependent. This study aimed to develop an integrated strategy based on the UPLC-DAD-QTOF-MS technique for metabolism and pharmacokinetic studies of HM. SL was used here as a test herb to verify the feasibility of the proposed strategy. SL was administered to rats, then, the blood plasma, urine and feces were analyzed to determine the metabolic profiles. Using our strategy, umbelliferone and scopoletin were evaluated to be the key bioactive components. Their pharmacokinetic parameters were measured and biotransformation pathways were elucidated. After oral administration of SL to rats, 17 components in blood, 10 components in urine and 2 components in feces were identified and characterized using our UPLC-DAD-QTOF-MS method. Umbelliferone, scopoletin and their metabolites were found to be the major components involved in the metabolism process. Literature reports also suggest that umbelliferone and scopoletin are responsible for the therapeutic effects of SL, thus these two components were selected as the active markers for pharmacokinetic study. In the test of validity, the established method presented good linearity with R(2)>0.99. The relative standard deviation value was below 13.9% for precision, and recovery studies for accuracy were found to be within the range 91.8-112.5%. The present strategy offers, simultaneously, precision in quantitative analysis (metabolism study) and accuracy in quantitative analysis (pharmacokinetic study) with greater efficiency and less costs, which is therefore reliably used for integrated metabolism and pharmacokinetic studies of HM.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Supelco
Umbelliferone, analytical standard
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Formic acid, reagent grade, ≥95%