Merck
  • Vascular heterogeneity between native rat pancreatic islets is responsible for differences in survival and revascularisation post transplantation.
Vascular heterogeneity between native rat pancreatic islets is responsible for differences in survival and revascularisation post transplantation.
Diabetologia (2014-09-27)
Sara Ullsten, Joey Lau, Per-Ola Carlsson
ABSTRACT
Highly blood-perfused islets have been observed to be the most functional islets in the native pancreas. We hypothesised that differences in vascular support of islets in donor pancreases influence their susceptibility to cellular stress and capacity for vascular engraftment after transplantation. Highly blood-perfused islets in rats were identified by injection of microspheres into the ascending aorta before islet isolation. Cell death was evaluated after in vitro cytokine or hypoxia exposure, and 2 days post transplantation. One month post transplantation, islet engraftment, including vascular density, blood perfusion and oxygen tension (pO2) in the tissue, was evaluated. Microsphere-containing islets had a similar frequency of cell death during standard culture conditions but increased cell death after exposure to cytokines and hypoxia in comparison with other islets. Two days after transplantation the percentage of apoptotic or necrotic cells was also higher in grafts of such islets and 1 month post transplantation these grafts were composed of substantially more connective tissue. Grafts of highly blood-perfused islets in the native pancreas regained a higher vascular density, blood perfusion and pO2 in comparison with grafts of other islets. Native islets that are highly blood-perfused regained this feature after transplantation, indicating a superior capacity for revascularisation and post-transplant function. However, the same group of islets was more vulnerable to different kinds of cellular stress, which limited their early survival post transplantation. Preferential death of these most active islets may contribute to the high number of islets needed to provide cure with islet transplantation.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
SKU
Pack Size
Availability
Price
Quantity
Supelco
D-(+)-Glucose, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen peroxide solution, 30 % (w/w), for inorganic trace analysis, for ultratrace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)