MilliporeSigma
  • Antibodies to early EBV, CMV, and HHV6 antigens in systemic lupus erythematosus patients.
Antibodies to early EBV, CMV, and HHV6 antigens in systemic lupus erythematosus patients.
Scandinavian journal of rheumatology (2015-01-07)
N S Rasmussen, A H Draborg, C T Nielsen, S Jacobsen, G Houen
ABSTRACT
We investigated the antibody levels against early antigens of Epstein-Barr virus (EBV), cytomegalovirus (CMV), and human herpesvirus 6 (HHV6) in systemic lupus erythematosus (SLE) patients and healthy controls, and further correlated these antibodies to haematology/biochemistry, serology, and disease activity measures. Immunoglobulin (Ig)M, IgG, and IgA levels against the DNA polymerase processivity factors of EBV, CMV, and HHV6, termed early antigen diffuse (EA/D), pp52, and p41, respectively, were determined in plasma samples from 77 SLE patients and 29 healthy controls by using enzyme-linked immunosorbent assays (ELISAs). IgM, IgG, and IgA levels against EBV EA/D, and IgG and IgA levels against CMV pp52, were significantly higher in SLE patients compared with healthy controls. Furthermore, EBV EA/D- and CMV pp52-directed IgG levels were inversely and positively associated, respectively, with lymphocyte counts in SLE patients. None of the findings seemed to be associated with use of immunosuppressive medication. Our results suggest strong, but opposite, associations of lytic EBV and CMV infections with SLE. The amplified humoral responses to EBV EA/D and CMV pp52 in our SLE patient cohort probably reflect aberrant control of EBV and CMV reactivation. However, reactivation of EBV appeared to correlate with lymphopenic manifestations in SLE patients whereas CMV reactivation seemed to correlate with increments in lymphocyte levels.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Supelco
Diethanolamine, analytical standard
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Trolamine impurity B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%