MilliporeSigma
  • SUMOylation determines turnover and localization of nephrin at the plasma membrane.
SUMOylation determines turnover and localization of nephrin at the plasma membrane.
Kidney international (2014-06-19)
Irini Tossidou, Erik Himmelseher, Beina Teng, Hermann Haller, Mario Schiffer
ABSTRACT
Podocyte effacement and the reformation of foot processes and slit diaphragms can be induced within minutes experimentally. Therefore, it seems likely that the slit diaphragm proteins underlie orchestrated recycling mechanisms under the control of posttranslational modifiers. One of these modifiers, SUMO (small ubiquitin-like modifier), is an ubiquitin-like protein with a 20% corresponding identity to ubiquitin. Modification by SUMOs to proteins on lysine residues can block the ubiquitination of the same site leading to the stabilization of the target protein. Here we found in vitro and in vivo that nephrin is a substrate modified by SUMO proteins thereby increasing its steady-state level and expression at the plasma membrane. A conversion of lysines to arginines at positions 1114 and 1224 of the intracellular tail of murine nephrin led to decreased stability of nephrin, decreased expression at the plasma membrane, and decreased PI3K/AKT signaling. Furthermore, treatment of podocytes with the SUMOylation inhibitor ginkgolic acid led to reduced membrane expression of nephrin. Similarly, the conversion of lysine to arginine at position 1100 of human nephrin caused decreased stability and expression at the plasma membrane. As SUMOylation is a reversible process, our results suggest that SUMOylation participates in the tight orchestration of nephrin turnover at the slit diaphragm.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Cycloheximide, PESTANAL®, analytical standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard